Learn More
Grainger Gage Pin Set ZZ Minus 1.30mm-4.98mm
Catalog No. 502752396
Encompass_Preferred
The Vermont Gage Class X Go Pin Gauge 0.8834 is a precision ground pin gauge used for measuring the size of holes, slots, and recesses. It has a diameter of 0.8834 inches, a tolerance of +/- 0.00005 inches, and is made of hardened stainless steel with a matte finish. The pin gauge has a knurled grip, carbide scribed lines, and chamfered measuring surfaces. It can be used as a no-go gauge and is traceable to NIST standards. The pin gauge is manufactured in the USA by Vermont Gage.
Catalog No. 50-275-2396 Supplier Grainger Supplier No. 141G83
The Vermont Gage Class X Go Pin Gauge 0.8834 is a precision ground pin gauge used for measuring the size of holes, slots, and recesses. It has a diameter of 0.8834 inches, a tolerance of +/- 0.00005 inches, and is made of hardened stainless steel with a matte finish. The pin gauge has a knurled grip, carbide scribed lines, and chamfered measuring surfaces. It can be used as a no-go gauge and is traceable to NIST standards. The pin gauge is manufactured in the USA by Vermont Gage.