Learn More
Grainger Gage Pin Set ZZ Minus 14.00mm-16.48mm
Catalog No. 502751851
Encompass_Preferred
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 9 pins with diameters that are 0.001 inches smaller than the nominal size stamped on each pin. The sizes range from 0.061 inches to 0.312 inches in increments of 0.025 inches. The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. This is a high quality pin set for precision measurement and inspection applications.
Catalog No. 50-275-1851 Supplier Grainger Supplier No. 141G86
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 9 pins with diameters that are 0.001 inches smaller than the nominal size stamped on each pin. The sizes range from 0.061 inches to 0.312 inches in increments of 0.025 inches. The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. This is a high quality pin set for precision measurement and inspection applications.