Learn More
Grainger Gage Pin Set ZZ Minus .2515 -.5005
Catalog No. 502752176
Encompass_Preferred
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 18 pins ranging from 0.4995" to 1.0005" in diameter, with a tolerance of +/- 0.00005". The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. This pin set allows for precise measurement and calibration for quality control applications.
Catalog No. 50-275-2176 Supplier Grainger Supplier No. 141G42
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 18 pins ranging from 0.4995" to 1.0005" in diameter, with a tolerance of +/- 0.00005". The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. This pin set allows for precise measurement and calibration for quality control applications.