Learn More
Grainger Gage Pin Set ZZ Minus .5015 -.6255
Catalog No. 502752584
Encompass_Preferred
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel gage pins for calibrating gages, checking hole locations, and setting micrometers. The set includes 9 pins with diameters that are 0.001" to 0.009" smaller than the nominal size stamped on each pin. The sizes included are 0.061", 0.081", 0.101", 0.201", 0.301", 0.401", 0.501", 0.601", and 0.701". The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and organization. This is a high quality gage pin set for precision measurement applications.
Catalog No. 50-275-2584 Supplier Grainger Supplier No. 141G43
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel gage pins for calibrating gages, checking hole locations, and setting micrometers. The set includes 9 pins with diameters that are 0.001" to 0.009" smaller than the nominal size stamped on each pin. The sizes included are 0.061", 0.081", 0.101", 0.201", 0.301", 0.401", 0.501", 0.601", and 0.701". The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and organization. This is a high quality gage pin set for precision measurement applications.