Learn More
Grainger Gage Pin Set ZZ Minus .6265 -.7505
Catalog No. 502752548
Encompass_Preferred
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 9 pins with diameters that are 0.0001 inches smaller than the nominal size stamped on each pin. The pins are made from hardened steel with a black oxide finish for corrosion resistance. Diameters range from 0.061 to 0.112 inches in increments of 0.001 inches. The pins come in a fitted plastic case for storage and organization. Overall this is a high quality pin gage set for precision measurement applications.
Catalog No. 50-275-2548 Supplier Grainger Supplier No. 141G44
The Grainger Approved Gage Pin Set ZZ Minus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 9 pins with diameters that are 0.0001 inches smaller than the nominal size stamped on each pin. The pins are made from hardened steel with a black oxide finish for corrosion resistance. Diameters range from 0.061 to 0.112 inches in increments of 0.001 inches. The pins come in a fitted plastic case for storage and organization. Overall this is a high quality pin gage set for precision measurement applications.