Learn More
Grainger Gage Pin Set ZZ Plus 1.31mm-4.99mm
Catalog No. 502752384
Encompass_Preferred
The Grainger Approved Gage Pin Set (141H02) is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 17 pins ranging from 0.100" to 1.000" in 0.025" increments, with a tolerance of +/- 0.0002". The pins have a Rockwell hardness of C60-C65 and feature a knurled grip area for easy handling. They are made in the USA and come packaged in a fitted plastic case for storage and organization. The pins can be used to calibrate dial indicators, micrometers, calipers, and other inspection tools.
Catalog No. 50-275-2384 Supplier Grainger Supplier No. 141H02
The Grainger Approved Gage Pin Set (141H02) is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 17 pins ranging from 0.100" to 1.000" in 0.025" increments, with a tolerance of +/- 0.0002". The pins have a Rockwell hardness of C60-C65 and feature a knurled grip area for easy handling. They are made in the USA and come packaged in a fitted plastic case for storage and organization. The pins can be used to calibrate dial indicators, micrometers, calipers, and other inspection tools.