Learn More
Grainger Gage Pin Set ZZ Plus 14.00mm-16.48mm
Catalog No. 502751852
Encompass_Preferred
The Grainger Approved Gage Pin Set (141G68) is a set of precision ground steel pins used for calibrating measuring instruments and setting up precision assemblies. The set includes 18 pins ranging from 0.4990" to 1.0000" in increments of 0.001". The pins are made from hardened and ground alloy steel with a Rockwell hardness of C60-C65. They feature a knurled grip area and come in a fitted plastic case for storage and organization. The gage pins are traceable to NIST standards and are accurate to +/- 0.00004". This high-quality pin set is ideal for calibration, inspection, and precision assembly applications.
Catalog No. 50-275-1852 Supplier Grainger Supplier No. 141G68
The Grainger Approved Gage Pin Set (141G68) is a set of precision ground steel pins used for calibrating measuring instruments and setting up precision assemblies. The set includes 18 pins ranging from 0.4990" to 1.0000" in increments of 0.001". The pins are made from hardened and ground alloy steel with a Rockwell hardness of C60-C65. They feature a knurled grip area and come in a fitted plastic case for storage and organization. The gage pins are traceable to NIST standards and are accurate to +/- 0.00004". This high-quality pin set is ideal for calibration, inspection, and precision assembly applications.