Learn More
Grainger Gage Pin Set ZZ Plus 16.50mm-18.98mm
Catalog No. 502752385
Encompass_Preferred
The Grainger Approved Gage Pin Set (141G69) is a set of precision ground steel pins used to calibrate measuring tools and instruments. The set includes 9 pins ranging from 0.100" to 1.000" in 0.100" increments, with a tolerance of +/- 0.0002". The pins have a Rockwell hardness of C60-C65 and feature a knurled grip area for handling. They are made in the USA and come packaged in a fitted plastic case for storage and organization. The pins allow for accurate calibration and inspection of gages, micrometers, dial indicators, and other precision measurement tools.
Catalog No. 50-275-2385 Supplier Grainger Supplier No. 141G69
The Grainger Approved Gage Pin Set (141G69) is a set of precision ground steel pins used to calibrate measuring tools and instruments. The set includes 9 pins ranging from 0.100" to 1.000" in 0.100" increments, with a tolerance of +/- 0.0002". The pins have a Rockwell hardness of C60-C65 and feature a knurled grip area for handling. They are made in the USA and come packaged in a fitted plastic case for storage and organization. The pins allow for accurate calibration and inspection of gages, micrometers, dial indicators, and other precision measurement tools.