Learn More
Grainger Gage Pin Set ZZ Plus .20mm-1.28mm
Catalog No. 502752486
Encompass_Preferred
The Grainger Approved Gage Pin Set - ZZ Plus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 22 pins ranging from 0.4990 to 1.0000 inches in diameter, with increments of 0.001 inches. The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. The pins are traceable to NIST standards for quality assurance. This pin set is an economical solution for basic dimensional gage calibration and inspection applications.
Catalog No. 50-275-2486 Supplier Grainger Supplier No. 141G64
The Grainger Approved Gage Pin Set - ZZ Plus is a set of precision ground steel pins used for calibrating gages, checking hole locations, and setting up inspection tools. The set contains 22 pins ranging from 0.4990 to 1.0000 inches in diameter, with increments of 0.001 inches. The pins are made from hardened steel with a black oxide finish for corrosion resistance. They come in a fitted plastic case for storage and portability. The pins are traceable to NIST standards for quality assurance. This pin set is an economical solution for basic dimensional gage calibration and inspection applications.