Learn More
Grainger Gage Pin Set ZZ Plus .21mm-1.29m
Catalog No. 502752488
Encompass_Preferred
The Grainger 141H01 is a 14 gauge, type 1 steel sheet metal that is 4 feet x 8 feet in size. It has a smooth finish and is 0.075 inches thick. The sheet metal is used for applications like ducts, furniture, and enclosures. It can be cut, formed, welded, and finished to specification. The product weighs approximately 49 pounds per sheet. Overall, the Grainger 141H01 is a versatile, strong, smooth sheet metal that can be fabricated for many uses.
Catalog No. 50-275-2488 Supplier Grainger Supplier No. 141H01
The Grainger 141H01 is a 14 gauge, type 1 steel sheet metal that is 4 feet x 8 feet in size. It has a smooth finish and is 0.075 inches thick. The sheet metal is used for applications like ducts, furniture, and enclosures. It can be cut, formed, welded, and finished to specification. The product weighs approximately 49 pounds per sheet. Overall, the Grainger 141H01 is a versatile, strong, smooth sheet metal that can be fabricated for many uses.