Learn More
Grainger Gage Pin Set ZZ Plus 5.00mm-9.98mm
Catalog No. 502752549
Encompass_Preferred
The Grainger Approved Gage Pin Set (141G66) is a set of steel gage pins used to calibrate measuring tools and instruments. The set includes 9 pins ranging from 0.4990 to 1.4995 inches in 0.001 inch increments. The pins are made from hardened and ground alloy steel with a matte chrome finish. They feature a knurled grip area and come in a fitted plastic case for storage and organization. The gage pins allow for precision calibration and are suitable for use in quality control departments, inspection areas, tool rooms and more.
Catalog No. 50-275-2549 Supplier Grainger Supplier No. 141G66
The Grainger Approved Gage Pin Set (141G66) is a set of steel gage pins used to calibrate measuring tools and instruments. The set includes 9 pins ranging from 0.4990 to 1.4995 inches in 0.001 inch increments. The pins are made from hardened and ground alloy steel with a matte chrome finish. They feature a knurled grip area and come in a fitted plastic case for storage and organization. The gage pins allow for precision calibration and are suitable for use in quality control departments, inspection areas, tool rooms and more.