Learn More
Grainger Gage Pn Set ZZ Plus .006 -.625 620 pcs.
Catalog No. 502751174
Encompass_Preferred
The Grainger ZZ-Plus Gage Pin Set contains 620 precision ground steel pins in 1/64" increments from 1/64" to 1". The pins are made from hardened and tempered alloy steel, precision ground within 0.0002" tolerance, and feature a knurled grip area for easy handling. This comprehensive set of pins is ideal for calibrating, setting, and checking tolerances, gaps, and clearances in inspection applications across a wide range of industries including manufacturing, automotive, aerospace, and more. The pins are housed in a sturdy plastic case with molded slots and a hinged lid for convenient storage and portability.
Catalog No. 50-275-1174 Supplier Grainger Supplier No. 141F68
The Grainger ZZ-Plus Gage Pin Set contains 620 precision ground steel pins in 1/64" increments from 1/64" to 1". The pins are made from hardened and tempered alloy steel, precision ground within 0.0002" tolerance, and feature a knurled grip area for easy handling. This comprehensive set of pins is ideal for calibrating, setting, and checking tolerances, gaps, and clearances in inspection applications across a wide range of industries including manufacturing, automotive, aerospace, and more. The pins are housed in a sturdy plastic case with molded slots and a hinged lid for convenient storage and portability.