Learn More
Aviva Systems Biology Corp GAGE2A Antibody - C-terminal region (ARP64957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAGE2A. It was validated on Western Blot. . Alternate names: CT4.2, GAGE2, GAGE2B, MGC96883, MGC96930, MGC96942ApplicationWB. Species reactivity: Human
Catalog No. 50-288-399