Learn More
Medchemexpress LLC GAK inhibitor 49 | 319492-82-5 | 98.8% | 370.40 g/mol | C20H22N2O5 | 100MG
SDP
Catalog No. 5000235321
Encompass_Preferred
GAK inhibitor 49 is a potent, ATP-competitive, and highly selective inhibitor of cyclin G-associated kinase (GAK) provided as a solid research compound. It exhibits low-nanomolar biochemical potency and cellular activity, making it suitable for probing GAK-dependent signaling in biochemical and cell-based studies. Soluble in DMSO and available as pre-made DMSO solutions for convenient use.
  • Potent GAK inhibition (Ki 0.54 nM).
  • Cellular activity with IC50 56 nM.
  • High purity, suitable for research use.
  • Molecular weight 370.40 g/mol.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-002-35321 Supplier Medchemexpress LLC Supplier No. HY124793100MG
GAK inhibitor 49 is a potent, ATP-competitive, and highly selective inhibitor of cyclin G-associated kinase (GAK) provided as a solid research compound. It exhibits low-nanomolar biochemical potency and cellular activity, making it suitable for probing GAK-dependent signaling in biochemical and cell-based studies. Soluble in DMSO and available as pre-made DMSO solutions for convenient use.
  • Potent GAK inhibition (Ki 0.54 nM).
  • Cellular activity with IC50 56 nM.
  • High purity, suitable for research use.
  • Molecular weight 370.40 g/mol.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Suitable for biochemical and cell-based assays.