Learn More
Medchemexpress LLC GAK inhibitor 49 hydrochloride | 2930378-91-7 | 99.5% | 406.86 | C20H23ClN2O5 | 10MG
SDP
Catalog No. 5000235614
Encompass_Preferred
GAK inhibitor 49 hydrochloride is the hydrochloride salt of a potent, ATP-competitive inhibitor of cyclin G-associated kinase (GAK) used for biochemical and cell-based research. It shows subnanomolar biochemical potency (Ki = 0.54 nM), low-nanomolar cellular activity (cell IC50 = 56 nM), and reported binding to RIPK2. Supplied as a high-purity solid for laboratory assays.
  • Potent ATP-competitive inhibition of cyclin G-associated kinase.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity typically ≥99.5% for research applications.
  • Suitable for biochemical kinase assays and cell-based target validation.
  • Available in milligram-scale quantities for research use.
Catalog No. 50-002-35614 Supplier Medchemexpress LLC Supplier No. HY124793A10MG
Add to Cart
Add to Cart
GAK inhibitor 49 hydrochloride is the hydrochloride salt of a potent, ATP-competitive inhibitor of cyclin G-associated kinase (GAK) used for biochemical and cell-based research. It shows subnanomolar biochemical potency (Ki = 0.54 nM), low-nanomolar cellular activity (cell IC50 = 56 nM), and reported binding to RIPK2. Supplied as a high-purity solid for laboratory assays.
  • Potent ATP-competitive inhibition of cyclin G-associated kinase.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity typically ≥99.5% for research applications.
  • Suitable for biochemical kinase assays and cell-based target validation.
  • Available in milligram-scale quantities for research use.