Learn More
Medchemexpress LLC GAK inhibitor 49 hydrochloride | 2930378-91-7 | 99.5% | 406.86 | C20H23ClN2O5 | 50MG
SDP
Catalog No. 5000236198
Encompass_Preferred
GAK inhibitor 49 hydrochloride is a potent, ATP-competitive, and selective cyclin G-associated kinase (GAK) inhibitor intended for biochemical and cellular research. Supplied as the hydrochloride salt in solid form, the material is provided in small pack sizes suitable for in vitro kinase assays, cellular inhibition studies, and mechanism-of-action experiments. Handle under sealed storage away from moisture; for solutions, store at -80°C for long-term stability.
  • Potent ATP-competitive inhibition of GAK.
  • Demonstrated cellular activity for target validation.
  • High purity (approximately 99.5%).
  • Provided as a solid hydrochloride salt for ease of handling.
  • Small pack suitable for research-scale experiments (50 mg).
  • Stable when stored sealed and kept moisture-free; long-term storage in solvent at -80°C.
Catalog No. 50-002-36198 Supplier Medchemexpress LLC Supplier No. HY124793A50MG
GAK inhibitor 49 hydrochloride is a potent, ATP-competitive, and selective cyclin G-associated kinase (GAK) inhibitor intended for biochemical and cellular research. Supplied as the hydrochloride salt in solid form, the material is provided in small pack sizes suitable for in vitro kinase assays, cellular inhibition studies, and mechanism-of-action experiments. Handle under sealed storage away from moisture; for solutions, store at -80°C for long-term stability.
  • Potent ATP-competitive inhibition of GAK.
  • Demonstrated cellular activity for target validation.
  • High purity (approximately 99.5%).
  • Provided as a solid hydrochloride salt for ease of handling.
  • Small pack suitable for research-scale experiments (50 mg).
  • Stable when stored sealed and kept moisture-free; long-term storage in solvent at -80°C.