Learn More
Medchemexpress LLC GAK inhibitor 49 hydrochloride | 2930378-91-7 | 99.5% | 406.86 g/mol | C20H23ClN2O5 | 25MG
SDP
Catalog No. 5000235154
Encompass_Preferred
GAK inhibitor 49 hydrochloride is the hydrochloride salt of a potent, ATP-competitive inhibitor of cyclin G-associated kinase used for biochemical and cell-based research. It exhibits subnanomolar binding affinity and cellular activity, suitable for kinase profiling and studies of GAK-related signaling.
  • Potent ATP-competitive inhibitor of cyclin G-associated kinase; Ki ≈ 0.54 nM.
  • Cellular activity with reported cell IC50 ≈ 56 nM.
  • High reported purity (≈99.5%) suitable for research applications.
  • Supplied as a solid (white to yellow) for easy handling and storage.
  • Recommended storage conditions and stability guidance for solids and solutions.
  • Available in small research quantities for assay development and profiling.
Catalog No. 50-002-35154 Supplier Medchemexpress LLC Supplier No. HY124793A25MG
GAK inhibitor 49 hydrochloride is the hydrochloride salt of a potent, ATP-competitive inhibitor of cyclin G-associated kinase used for biochemical and cell-based research. It exhibits subnanomolar binding affinity and cellular activity, suitable for kinase profiling and studies of GAK-related signaling.
  • Potent ATP-competitive inhibitor of cyclin G-associated kinase; Ki ≈ 0.54 nM.
  • Cellular activity with reported cell IC50 ≈ 56 nM.
  • High reported purity (≈99.5%) suitable for research applications.
  • Supplied as a solid (white to yellow) for easy handling and storage.
  • Recommended storage conditions and stability guidance for solids and solutions.
  • Available in small research quantities for assay development and profiling.