Learn More
Bon Opus Biosciences Recombinant Human ERVK-7 Protein(C-His) - Source: E. coli
SDP
Catalog No. NC2733729
Encompass_Preferred
Recombinant Human ERVK-7 Protein is produced by E.coli expression system and the target gene encoding G2-L130 is expressed with a His tag at the C-terminus. Accession#P63130. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733729 Supplier Bon Opus Biosciences Supplier No. BP251850UG
Add to Cart
Add to Cart
Recombinant Human ERVK-7 Protein is produced by E.coli expression system and the target gene encoding G2-L130 is expressed with a His tag at the C-terminus. Accession#P63130. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4