Learn More
Medchemexpress LLC GAL-02-221 | 2409538-70-9 | 99.9% | 1016.67 | 10 MG
SDP
Catalog No. 5000365108
Encompass_Preferred
GAL-02-221 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4. It can potently degrade BRD4 in HER2 positive and negative breast cancer cell lines.
  • Potently degrades BRD4
  • Degrades BRD4 in HER2 positive and negative breast cancer cell lines
  • Connected by ligands for von Hippel-Lindau and BRD4
  • Shows antiproliferative activity against human 22Rv1, L02, MCF7, and MDA-MB-231 cells
Catalog No. 50-003-65108 Supplier Medchemexpress LLC Supplier No. 10MGHY133737
GAL-02-221 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4. It can potently degrade BRD4 in HER2 positive and negative breast cancer cell lines.
  • Potently degrades BRD4
  • Degrades BRD4 in HER2 positive and negative breast cancer cell lines
  • Connected by ligands for von Hippel-Lindau and BRD4
  • Shows antiproliferative activity against human 22Rv1, L02, MCF7, and MDA-MB-231 cells