Learn More
Medchemexpress LLC HY-101422 5mg Medchemexpress, GAL-021 CAS:1380341-99-0 Purity:>98%
SDP
Catalog No. 501932121
Encompass_Preferred
Medchemexpress, HY-101422 5mg GAL-021 CAS:1380341-99-0 GAL-021 a new intravenous BKCa-channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2121 Supplier Medchemexpress LLC Supplier No. HY1014225MG
Medchemexpress, HY-101422 5mg GAL-021 CAS:1380341-99-0 GAL-021 a new intravenous BKCa-channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.