Learn More
TARGETMOL CHEMICALS INC GAL-021 sulfate 5MG
SDP
Catalog No. 502880006
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GAL-021 sulfate is a BKCa channel blocker that inhibits the analgesic effects of opioids and is used in the study of respiratory control diseases. Purity 99.68%
Catalog No. 50-288-0006 Supplier TARGETMOL CHEMICALS INC Supplier No. T612395MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GAL-021 sulfate is a BKCa channel blocker that inhibits the analgesic effects of opioids and is used in the study of respiratory control diseases. Purity 99.68%