Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP 92.0+%, TCI America™
SDP

Catalog No. M17265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17265G 5 g
M17261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17265G Supplier TCI America Supplier No. M17265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 150412-81-0 | C61H64O12 | 989.171 g/mol

Chemical Identifiers

CAS 150412-81-0
Molecular Formula C61H64O12
Molecular Weight (g/mol) 989.171
InChI Key GEVBSODSBYNKQA-IMBADGMBSA-N
PubChem CID 11320628
IUPAC Name (2R,3S,4S,5R,6S)-6-[(2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-yl]oxy-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OC4C(C(C(C(O4)COCC5=CC=CC=C5)O)OCC6=CC=CC=C6)OCC7=CC=CC=C7)OCC8=CC=CC=C8)OCC9=CC=CC=C9

Specifications

Color White
Quantity 5 g
Synonym LacMP 4′-OH, per OBn, 4-Methoxyphenyl 4-O-(2,3,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside
Formula Weight 989.17
Percent Purity ≥92.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.