Learn More
American Radiolabeled Chemicals Inc GAL D-1-14C 1 PO4 50UC
SDP
Catalog No. 50901280
Encompass_Preferred
Galactose, D-1-14C 1-phosphate 50uCi in ethanol:water (7:3), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 294, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-280 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 347350UC
Galactose, D-1-14C 1-phosphate 50uCi in ethanol:water (7:3), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 294, Shipped in Dry Ice, Exclusive