Learn More
Aviva Systems Biology Corp GAL3ST3 Antibody - middle region (ARP50150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAL3ST3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GAL3ST3. Alternate names: GAL3ST-3, GAL3ST2, MGC142112, MGC142114ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-494