Learn More
Aviva Systems Biology Corp Gal3st4 Antibody - N-terminal region (ARP49843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gal3st4. It was validated on Western Blot. . Alternate names: RGD1561105ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-456