Learn More
BIOHIPPO GAL4-TAL-BSD LV
SDP
Catalog No. 5000331672
Encompass
GAL4-TAL-BSD is a transcription factor reporter lentiviral vector designed to monitor transcription factor pathways in living cells This construct delivers a Firefly Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Identification of protein-protein interactions (PPI) and as a reporter in mammalian two-hybrid assay The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31672 Supplier BIOHIPPO Supplier No. BHV194000344
GAL4-TAL-BSD is a transcription factor reporter lentiviral vector designed to monitor transcription factor pathways in living cells This construct delivers a Firefly Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Identification of protein-protein interactions (PPI) and as a reporter in mammalian two-hybrid assay The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures