Learn More
Medchemexpress LLC Gala Tfa | 99.3% | 3032.36 | 10 MG
SDP
Catalog No. 5000399612
Encompass_Preferred
GALA (TFA) is a biologically active peptide that is pH-sensitive and fusogenic. It is a promising method to accelerate the process of enhancing the expression of desired proteins. This product is for research use only.
  • Biologically active peptide
  • pH-sensitive and fusogenic
  • Enhances expression of desired proteins
  • For research use only
Catalog No. 50-003-99612 Supplier Medchemexpress LLC Supplier No. HYP5423A10MG
GALA (TFA) is a biologically active peptide that is pH-sensitive and fusogenic. It is a promising method to accelerate the process of enhancing the expression of desired proteins. This product is for research use only.
  • Biologically active peptide
  • pH-sensitive and fusogenic
  • Enhances expression of desired proteins
  • For research use only