Learn More
Medchemexpress LLC GALA (TFA) | 99.3% | 3032.36 | 5 MG
SDP
Catalog No. 5000399615
Encompass_Preferred
GALA (TFA) is a biologically active, pH-sensitive fusogenic peptide. It is considered a promising method to accelerate protein expression and enhance the transfection efficiency of cationic liposomes. This product is intended for research use only.
  • Biologically active peptide
  • pH-sensitive fusogenic peptide
  • Enhances expression of desired proteins
  • Enhances transfection efficiency of cationic liposomes
  • For research use only
Catalog No. 50-003-99615 Supplier Medchemexpress LLC Supplier No. HYP54235MG
GALA (TFA) is a biologically active, pH-sensitive fusogenic peptide. It is considered a promising method to accelerate protein expression and enhance the transfection efficiency of cationic liposomes. This product is intended for research use only.
  • Biologically active peptide
  • pH-sensitive fusogenic peptide
  • Enhances expression of desired proteins
  • Enhances transfection efficiency of cationic liposomes
  • For research use only