Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gala tfa | 99.3% | 3032.36 (free base) | 1 MG
SDP

Catalog No. 5000398379
Encompass_Preferred

GALA (TFA) is a biologically active peptide and a pH-sensitive fusogenic peptide. It is a promising method to accelerate the process of enhancing the expression of desired proteins.

  • Purity: 99.34%
  • Molecular weight: 3032.36 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Trp-Glu-Ala-Ala-Leu-Ala-Glu-Ala-Leu-Ala-Glu-Ala-Leu-Ala-Glu-His-Leu-Ala-Glu-Ala-Leu-Ala-Glu-Ala-Leu-Glu-Ala-Leu-Ala-Ala
  • Solubility (in vitro): DMSO: 12.5 mg/mL (ultrasonic and warming and heat to 60°C)

Catalog No. 50-003-98379 Supplier Medchemexpress LLC Supplier No. HYP5423A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GALA (TFA) is a biologically active peptide and a pH-sensitive fusogenic peptide. It is a promising method to accelerate the process of enhancing the expression of desired proteins.

  • Purity: 99.34%
  • Molecular weight: 3032.36 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Trp-Glu-Ala-Ala-Leu-Ala-Glu-Ala-Leu-Ala-Glu-Ala-Leu-Ala-Glu-His-Leu-Ala-Glu-Ala-Leu-Ala-Glu-Ala-Leu-Glu-Ala-Leu-Ala-Ala
  • Solubility (in vitro): DMSO: 12.5 mg/mL (ultrasonic and warming and heat to 60°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.