Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GALA TFA | 99.3% | 3032.36 (free base) | 5 MG
SDP

Catalog No. 5000399613
Encompass_Preferred

GALA (TFA) is a biologically active peptide, a pH-sensitive fusogenic peptide that can accelerate the process of enhancing the expression of desired proteins. It is intended for research use only and not for patient administration.

  • Purity: 99.34%
  • Molecular weight: 3032.36 (free base)
  • Appearance: Solid, white to off-white
  • Storage: Powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month. Sealed, away from moisture and light.
  • Solubility (in vitro): 12.5 mg/mL in DMSO (with ultrasonic, warming, and heating to 60°C).
  • Related classifications: Peptides, peptide derivatives, peptides for drug delivery, and responsive peptides.

Catalog No. 50-003-99613 Supplier Medchemexpress LLC Supplier No. HYP5423A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GALA (TFA) is a biologically active peptide, a pH-sensitive fusogenic peptide that can accelerate the process of enhancing the expression of desired proteins. It is intended for research use only and not for patient administration.

  • Purity: 99.34%
  • Molecular weight: 3032.36 (free base)
  • Appearance: Solid, white to off-white
  • Storage: Powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month. Sealed, away from moisture and light.
  • Solubility (in vitro): 12.5 mg/mL in DMSO (with ultrasonic, warming, and heating to 60°C).
  • Related classifications: Peptides, peptide derivatives, peptides for drug delivery, and responsive peptides.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.