Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galactan, ex gum arabic, Thermo Scientific™

Catalog No. AC225891000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC225891000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225891000 Supplier Thermo Scientific Chemicals Supplier No. 225891000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39300-87-3
Infrared Spectrum Authentic
Molecular Formula C20H36O16
Quantity 100g
MDL Number MFCD00134651
Synonym methyl 4-o-methylhexopyranosyl-1->4 hexopyranosyl-1->4 hexopyranoside, 5-5-3,4-dihydroxy-6-hydroxymethyl-5-methoxyoxan-2-yl oxy-3,4-dihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-hydroxymethyl-2-methoxyoxane-3,4-diol
InChI Key HYRUXGRHTJRKNG-UHFFFAOYSA-N
SMILES COC1C(OC(C(C1O)O)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)CO
IUPAC Name 2-[6-[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular Weight (g/mol) 532.492
PubChem CID 20758205
Chemical Name or Material Galactan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.