Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galactinol Hydrate 99.0+%, TCI America™
SDP

Catalog No. G0298100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0298100MG 100 mg
G02981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0298100MG Supplier TCI America Supplier No. G0298100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3687-64-7
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.327
InChI Key HGCURVXTXVAIIR-FMQVHOFPSA-N
Synonym 1-O-alpha-D-Galactopyranosyl-L-myo-inositol
PubChem CID 45052268
IUPAC Name (1S,2R,4S,5S)-6-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxycyclohexane-1,2,3,4,5-pentol;dihydrate
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(C2O)O)O)O)O)O)O)O)O.O.O

Specifications

Melting Point 221°C
Color White
Quantity 100 mg
Formula Weight 342.30
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Galactinol Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.