Learn More
Cayman Chemical Galactinol (hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502968076
Encompass_Preferred
An α-galactoside of inositol that serves as a galactosyl donor in the transglycosidase-catalyzed biosynthesis of the raffinose family; generated from myo-inositol and uridine diphosphate galactose in a galactosyltransferase-catalyzed reaction
Catalog No. 50-296-8076 Supplier Cayman Chemical Supplier No. 18426100 MG
An α-galactoside of inositol that serves as a galactosyl donor in the transglycosidase-catalyzed biosynthesis of the raffinose family; generated from myo-inositol and uridine diphosphate galactose in a galactosyltransferase-catalyzed reaction