Learn More
Cayman Chemical GalactInolhydrate 10mg
SDP
Catalog No. 502965731
Encompass_Preferred
An α-galactoside of inositol that serves as a galactosyl donor in the transglycosidase-catalyzed biosynthesis of the raffinose family; generated from myo-inositol and uridine diphosphate galactose in a galactosyltransferase-catalyzed reaction
Catalog No. 50-296-5731 Supplier Cayman Chemical Supplier No. 1842610 MG
An α-galactoside of inositol that serves as a galactosyl donor in the transglycosidase-catalyzed biosynthesis of the raffinose family; generated from myo-inositol and uridine diphosphate galactose in a galactosyltransferase-catalyzed reaction