Learn More
Cayman Chemical galacto-DapaglIflozIn 50mg
SDP
Catalog No. 502972090
Encompass_Preferred
A potent inhibitor of human SGLT2 that is 1,000-fold less potent at human SGLT1 (Ki values are 2 and 25,000 nM, respectively); dissociates from hSGLT2 more slowly than from hSGLT1 (half-time off rates are 19 and 0.9 seconds, respectively)
Catalog No. 50-297-2090 Supplier Cayman Chemical Supplier No. 1595250 MG
A potent inhibitor of human SGLT2 that is 1,000-fold less potent at human SGLT1 (Ki values are 2 and 25,000 nM, respectively); dissociates from hSGLT2 more slowly than from hSGLT1 (half-time off rates are 19 and 0.9 seconds, respectively)