Learn More
Carbosynth LLC. Galacto-RGD2, 5mg, MW: (M.W. 2149.24), Molecular formula: C91H137N29O32
SDP
Supplier:  Carbosynth LLC. RGD3782PI5MG
Encompass_Preferred
Application: Galacto RGD dimer, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: S. Ji, A. Czerwinski, Y. Zhou, Gl Shao, F. Valenzuela, P. Sowinski, S. Chauhan, M. Pennington, and S. Liu, Mol. Pharmaceutics, 10, 3304 (2013).
Catalog No. 50-168-6946