Learn More
Medchemexpress LLC Galactose 1-phosphate-13C-1 potassium | 478518-80-8 | MFCD00070670 | 99.2% | 337.31 g/mol | C5 13CH11K2O9P | 5 MG
SDP
Catalog No. 5000262977
Encompass_Preferred
13C-labeled galactose 1-phosphate potassium is the dipotassium salt of galactose 1-phosphate labeled with 13C at the C-1 position. Supplied for research use, it serves as an isotopic tracer or analytical standard in studies of galactose metabolism and nucleotide-sugar pathways. The material is provided as a stable potassium salt with a molecular weight of 337.31 g/mol.
  • Isotopically labeled at the C-1 position for metabolic tracing and quantitation.
  • Potassium salt form, compatible with aqueous buffers and enzymatic assays.
  • High purity suitable for use as an analytical standard in mass spectrometry.
  • Stable solid form for reproducible handling and storage.
Catalog No. 50-002-62977 Supplier Medchemexpress LLC Supplier No. HY113143AS15MG
13C-labeled galactose 1-phosphate potassium is the dipotassium salt of galactose 1-phosphate labeled with 13C at the C-1 position. Supplied for research use, it serves as an isotopic tracer or analytical standard in studies of galactose metabolism and nucleotide-sugar pathways. The material is provided as a stable potassium salt with a molecular weight of 337.31 g/mol.
  • Isotopically labeled at the C-1 position for metabolic tracing and quantitation.
  • Potassium salt form, compatible with aqueous buffers and enzymatic assays.
  • High purity suitable for use as an analytical standard in mass spectrometry.
  • Stable solid form for reproducible handling and storage.