Learn More
Medchemexpress LLC Galactose 1-phosphate-13C potassium | 478518-78-4 | 98.8% | 337.31 | 10 MG
SDP
Catalog No. 5000436478
Encompass_Preferred
Galactose 1-phosphate-13C (potassium) is the 13C labeled Galactose 1-phosphate Potassium salt, an intermediate in galactose metabolism and nucleotide sugar. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36478 Supplier Medchemexpress LLC Supplier No. HY113143AS10MG
Galactose 1-phosphate-13C (potassium) is the 13C labeled Galactose 1-phosphate Potassium salt, an intermediate in galactose metabolism and nucleotide sugar. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.