Learn More
Medchemexpress LLC Galactose 1-phosphate-¹³C-1 potassium | 478518-80-8 | 337.31 | 25 MG
SDP
Catalog No. 5000439106
Encompass_Preferred
Galactose 1-phosphate-¹³C-1 (potassium) is the ¹³C labeled Galactose 1-phosphate Potassium salt. Galactose 1-phosphate Potassium salt is an intermediate in the galactose metabolism and nucleotide sugar. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Intermediate in galactose metabolism and nucleotide sugar
  • Suitable for tracer applications in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-39106 Supplier Medchemexpress LLC Supplier No. 25MGHY113143AS1
Galactose 1-phosphate-¹³C-1 (potassium) is the ¹³C labeled Galactose 1-phosphate Potassium salt. Galactose 1-phosphate Potassium salt is an intermediate in the galactose metabolism and nucleotide sugar. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Intermediate in galactose metabolism and nucleotide sugar
  • Suitable for tracer applications in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs