Learn More
Medchemexpress LLC HY-112617A 100mg Medchemexpress, Thalidomide-O-amido-PEG2-C2-NH2 (TFA) CAS:1957235-75-4 Purity:>98%
SDP
Catalog No. 501969118
Encompass_Preferred
Medchemexpress, HY-113143A 50mg Galactose 1-phosphate Potassium salt CAS:19046-60-7 Galactose 1-phosphate Potassium salt is is an intermediate in the galactose metabolism and nucleotide sugars. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9118 Supplier Medchemexpress LLC Supplier No. HY112617A100MG
Medchemexpress, HY-113143A 50mg Galactose 1-phosphate Potassium salt CAS:19046-60-7 Galactose 1-phosphate Potassium salt is is an intermediate in the galactose metabolism and nucleotide sugars. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.