Learn More
Medchemexpress LLC Galactose 1-phosphate potassium salt | 19046-60-7 | MFCD00150979 | 99.0% | 336.32 g/mol | C6H11K2O9P | 5 MG
SDP
Catalog No. 5000264047
Encompass_Preferred
Galactose 1-phosphate potassium salt is a research-grade metabolite and intermediate in galactose metabolism and nucleotide sugar pathways. Supplied as a solid with defined purity and formulation options, it is intended for enzymology, biochemical assays, and metabolomics applications.
  • Intermediate in galactose metabolism and nucleotide sugar biosynthesis.
  • High purity (99.0%) for reliable experimental results.
  • White to off-white solid, suitable for handling and storage.
  • Available in small milligram quantities for lab-scale experiments.
  • Also available as a 10 mM solution in water for immediate use.
  • Suitable for enzymatic assays, metabolite profiling, and biochemical research.
Catalog No. 50-002-64047 Supplier Medchemexpress LLC Supplier No. HY113143A5MG
Galactose 1-phosphate potassium salt is a research-grade metabolite and intermediate in galactose metabolism and nucleotide sugar pathways. Supplied as a solid with defined purity and formulation options, it is intended for enzymology, biochemical assays, and metabolomics applications.
  • Intermediate in galactose metabolism and nucleotide sugar biosynthesis.
  • High purity (99.0%) for reliable experimental results.
  • White to off-white solid, suitable for handling and storage.
  • Available in small milligram quantities for lab-scale experiments.
  • Also available as a 10 mM solution in water for immediate use.
  • Suitable for enzymatic assays, metabolite profiling, and biochemical research.