Learn More
MOLECULAR DEPOT LLC Galactose - 500 mg
Catalog No. 5000013496
Encompass_Preferred
Galactose for Interference Testing is a simple monosaccharide that can be used as an energy source as well as a component of glycolipids and glycoproteins This product can used to test galactose interference in immunoassays For in vitro research use only Not for human or animal use
Catalog No. 50-000-13496 Supplier MOLECULAR DEPOT LLC Supplier No. C2010416
Galactose for Interference Testing is a simple monosaccharide that can be used as an energy source as well as a component of glycolipids and glycoproteins This product can used to test galactose interference in immunoassays For in vitro research use only Not for human or animal use