Learn More
Vector Laboratories Glycoprotein Eluting Solution for Galactose/GalNAc Binding Lectins | 100mL
SDP
Supplier:  Vector Laboratories ES2100100
Encompass
Convenient, ready-to-use solutions. Superior recovery over standard sugar solutions. Maximize the yield of eluted glycoproteins. Preserve the activity of the agarose-bound lectins for repeated use. Elution in neutral pH range. All components can be subsequently removed by dialysis.
Catalog No. NC1777775