Learn More
Medchemexpress LLC Galactose oxidase, Dactylium dendroides (GOase) | 9028-79-9 | ≥100 U/mg protein | ~68.5 kDa | 150 U
SDP
Catalog No. 5000400297
Encompass_Preferred
Galactose Oxidase, Dactylium dendroides (GOase) is a type II copper metalloenzyme derived from fungus, containing a single polypeptide. It is widely utilized in biochemical research due to its catalytic activity. This enzyme facilitates the two-electron oxidation of primary alcohols, converting them into their corresponding aldehydes, and concurrently reduces dioxygen to hydrogen peroxide.
  • Catalyzes two-electron oxidation of primary alcohols.
  • Reduces dioxygen to hydrogen peroxide.
  • Soluble in pure water (1 mg/mL).
  • Recommended buffer for catalytic reaction is 0.1 M Potassium phosphate buffer, pH 6.0.
Catalog No. 50-004-00297 Supplier Medchemexpress LLC Supplier No. HYP2910150U
Galactose Oxidase, Dactylium dendroides (GOase) is a type II copper metalloenzyme derived from fungus, containing a single polypeptide. It is widely utilized in biochemical research due to its catalytic activity. This enzyme facilitates the two-electron oxidation of primary alcohols, converting them into their corresponding aldehydes, and concurrently reduces dioxygen to hydrogen peroxide.
  • Catalyzes two-electron oxidation of primary alcohols.
  • Reduces dioxygen to hydrogen peroxide.
  • Soluble in pure water (1 mg/mL).
  • Recommended buffer for catalytic reaction is 0.1 M Potassium phosphate buffer, pH 6.0.