Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Galactose Oxidase, Dactylium dendroides (GOase) | 9028-79-9 | ~68.5 kDa | 50 U
SDP

Catalog No. 5000399866
Encompass_Preferred

Galactose Oxidase, Dactylium dendroides (GOase) is a type II copper metalloenzyme derived from fungus, commonly used in biochemical studies. It catalyzes the two-electron oxidation of primary alcohols to their corresponding aldehydes, reducing dioxygen to hydrogen peroxide.

  • Single polypeptide chain
  • Molecular weight of approximately 68.5 kDa
  • Specific activity of ≥100 U/mg protein
  • Soluble in pure water (1 mg/mL)
  • Catalytic reaction system buffer is recommended to be 0.1 M potassium phosphate buffer, pH 6.0
  • White to off-white solid appearance
  • Inhibited by cyanide, diethyldithiocarbamate, azide, hydroxylamine, EDTA

Catalog No. 50-003-99866 Supplier Medchemexpress LLC Supplier No. HYP291050U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Galactose Oxidase, Dactylium dendroides (GOase) is a type II copper metalloenzyme derived from fungus, commonly used in biochemical studies. It catalyzes the two-electron oxidation of primary alcohols to their corresponding aldehydes, reducing dioxygen to hydrogen peroxide.

  • Single polypeptide chain
  • Molecular weight of approximately 68.5 kDa
  • Specific activity of ≥100 U/mg protein
  • Soluble in pure water (1 mg/mL)
  • Catalytic reaction system buffer is recommended to be 0.1 M potassium phosphate buffer, pH 6.0
  • White to off-white solid appearance
  • Inhibited by cyanide, diethyldithiocarbamate, azide, hydroxylamine, EDTA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.