Learn More
Bioworld GALACTOSE SEPAROPOREAGARO
Catalog No. 50197686
Encompass
GALACTOSE SEPAROPORE (AGAROSE) 4B-CL: This product is composed of a Galactose molecule coupled with high strength bioagarose matrix, Separopore 4B-CL. Technical Specifications: Ligand: D-Galactose; Ligand coupling method: Covalent lin...
Catalog No. 50-197-686 Supplier Bioworld Supplier No. 201810112
GALACTOSE SEPAROPORE (AGAROSE) 4B-CL: This product is composed of a Galactose molecule coupled with high strength bioagarose matrix, Separopore 4B-CL. Technical Specifications: Ligand: D-Galactose; Ligand coupling method: Covalent lin...