Learn More
Roche Diagnostics Galactose/Arabinose Bio, 4 x 50 tests
Supplier:  Roche Diagnostics 08391599001
Encompass
This product is intended for quantitative determination of galactose or arabinose in aqueous solutions using the Cedex Bio Analyzer.
Catalog No. NC1524404