Learn More
Biomatik Corporation Galactosidase Alpha (Gla) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29212
Encompass_Preferred
Galactosidase Alpha (Gla) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Galactosidase Alpha (GLa). The Galactosidase Alpha (Gla) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Gly80. Applications: WB, IHC. Target Synonyms: Galactosidase Alpha (GLa). Accession Number: P06280. Gene ID: 2717. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4820