Learn More
Medchemexpress LLC β-Galactosidase, E. coli | 9031-11-2 | ≥50U/mg solid | 5 KU
SDP
Catalog No. 5000398633
Encompass_Preferred
β-Galactosidase (EC 3.2.1.23) is a glycoside hydrolase that breaks down β-glycosidic bonds between galactose and its organic parts. In E. coli, the lacZ gene is the structural gene for β-galactosidase, forming part of the lac operon's induction system. This enzyme can hydrolyze lactose into glucose and galactose, which then enter glycolysis.
  • Can catalyze the transgalactosylation of lactose into allolactose
  • Allolactose can further be broken into monosaccharides
Catalog No. 50-003-98633 Supplier Medchemexpress LLC Supplier No. HYP28695KU
β-Galactosidase (EC 3.2.1.23) is a glycoside hydrolase that breaks down β-glycosidic bonds between galactose and its organic parts. In E. coli, the lacZ gene is the structural gene for β-galactosidase, forming part of the lac operon's induction system. This enzyme can hydrolyze lactose into glucose and galactose, which then enter glycolysis.
  • Can catalyze the transgalactosylation of lactose into allolactose
  • Allolactose can further be broken into monosaccharides