Learn More
Apexbio Technology LLC β-Galactosidase Reporter Assay Kit 200T
SDP
Catalog No. 5000097199
Encompass_Preferred
Beta-Galactosidase Reporter Assay Kit is a colorimetric assay system designed to quantify gene expression by measuring -galactosidase enzyme activity It facilitates the evaluation of transcriptional activity in biological samples Beta-Galactosidase Reporter Assay Kit exerts its function by enabling the -galactosidase-mediated hydrolysis of ortho-nitrophenyl- -D-galactopyranoside (ONPG) generating a yellow product detectable at 420 nm In cell-based studies this assay is commonly utilized alongside luciferase-based systems for dual-reporter experiments to normalize for transfection efficiency Based on these features Beta-Galactosidase Reporter Assay Kit is broadly applicable in gene expression analysis signal transduction research and studies requiring normalization controls
Catalog No. 50-000-97199 Supplier Apexbio Technology LLC Supplier No. K2183200
Beta-Galactosidase Reporter Assay Kit is a colorimetric assay system designed to quantify gene expression by measuring -galactosidase enzyme activity It facilitates the evaluation of transcriptional activity in biological samples Beta-Galactosidase Reporter Assay Kit exerts its function by enabling the -galactosidase-mediated hydrolysis of ortho-nitrophenyl- -D-galactopyranoside (ONPG) generating a yellow product detectable at 420 nm In cell-based studies this assay is commonly utilized alongside luciferase-based systems for dual-reporter experiments to normalize for transfection efficiency Based on these features Beta-Galactosidase Reporter Assay Kit is broadly applicable in gene expression analysis signal transduction research and studies requiring normalization controls